Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cleanroom tapes, PCX™

Cleanroom tapes, PCX™

Cleanroom tape, White, -, 25,4 mm
Catalog # 819-0031
Cleanroom tapes, PCX™
Catalog # 819-0031
Supplier:  
Cleanroom tapes, PCX™
Cleanroom tapes, PCX™

Cleanroom tape, White, -, 25,4 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
TAPE PCX PE 1INX36YDS WHITE 1 * 36 Roll Catalog #MICRPCX-1WH

Product MICRPCX-1WH is a replacement for 819-0031 but varies by Brand, Packaging or Quantity.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These cleanroom tapes are made of LDPE with acrylic adhesive.

  • Labeling
  • Sealing, wafer box sealing
  • Over labeling (protect paper labels in water baths)

A low density polyethylene tape, low in halogens and leachable chlorides. Processed and packaged in a cleanroom. The polyethylene film and acrylic adhesive lessen the chance of outgassing. PCX offers excellent chemical resistance and good conformability. Works well with polycarbonates, glass and acrylics. Suitable for the following:

Length: 32,9 m

Different colours available.

Specifications
  • Sterile:
    -
  • Width:
    25,4 mm
  • Colour:
    White
  • Description:
    Cleanroom tape
  • Seq:
    11

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.