Single channel micro pipettes, electronic, variable / fixed volume, Accumax VA-FA
50 µl, Blue
Catalog # FINEFA-0050
Supplier: ACCUMAX
Single channel micro pipettes, electronic, variable / fixed volume, Accumax VA-FA
Catalog # FINEFA-0050
Supplier: ACCUMAX
Jump to:
Product Details & Documents
These pipettors feature a click-stop digital system for volume setting which ensures accurate dispensing of the liquid. The two-step plunger operation allows a reverse pipetting technique.
- Smooth plunger mechanism for ease of operation
- Tip ejector for smooth ejection of the tip preventing any contamination
- Ergonomic, lightweight design with comfortable finger rest
- Durable tip cone offers high resistance to shocks and chemical corrosion
- Colour-coded for easy identification
- Autoclavable (lower part)
Ordering information: Supplied with calibration report.
Specifications
- Colour code:Blue
- Volume:50 µl
- Seq:14