Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Microscope slides

Microscope slides

Microscope slides with one cavity, 1,0 - 1,2 mm, 76×26 mm
Catalog # SMITMC2952B/76X26
Supplier:   SMITH SCIENTIFIC
Microscope slides
Catalog # SMITMC2952B/76X26
Supplier:  SMITH SCIENTIFIC
Microscope slides

Microscope slides with one cavity, 1,0 - 1,2 mm, 76×26 mm

Price
£89.70
Log in for availability
1 * 50 items
Product Details & Documents

These slides are made of glass with ground edges.

Specifications
  • Description:
    Microscope slides with one cavity
  • L×W:
    76×26 mm
  • Seq:
    005
  • Thickness:
    1,0 - 1,2 mm