Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Microscope slides

Microscope slides

Microscope slides, 1,0 - 1,2 mm, 76×38 mm
Catalog # SMITMC2951B/76X38
Supplier:   SMITH SCIENTIFIC
Microscope slides
Catalog # SMITMC2951B/76X38
Supplier:  SMITH SCIENTIFIC
Microscope slides

Microscope slides, 1,0 - 1,2 mm, 76×38 mm

Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

These slides are made of glass with ground edges.

Specifications
  • Description:
    Microscope slides
  • L×W:
    76×38 mm
  • Seq:
    002
  • Thickness:
    1,0 - 1,2 mm