Volumetric flasks, class B
200 ml, 14/23, 0,3 ± m
Catalog # SMIT1503B-200
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
Borosilicate glass, with plastic stopper.
Certifications: ISO 1042
Specifications
- NS:14/23
- Capacity:200 ml
- Seq:04
- Tolerance:0,3 ± m