Volumetric flasks, class B
10 ml, 7/16, 0,05 ± m
Catalog # SMIT1503B-10
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
Borosilicate glass, with plastic stopper.
Certifications: ISO 1042
Specifications
- NS:7/16
- Capacity:10 ml
- Seq:01
- Tolerance:0,05 ± m