Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sub blotting system, omniPAGE

Sub blotting system, omniPAGE

Mini-wide blotting system, 100×200 mm
Catalog # CLEVSB10W
Supplier:   CLEAVER SCIENTIFIC
Sub blotting system, omniPAGE
Catalog # CLEVSB10W
Supplier:  CLEAVER SCIENTIFIC
Sub blotting system, omniPAGE
Sub blotting system, omniPAGE

Mini-wide blotting system, 100×200 mm

Price
£1,420.00
Log in for availability
1 * 1 items
Product Details & Documents

Each omniPAGE system comprises an omniPAGE tank and lid, electroblotting insert, cassettes and fibre pads. The versatile design of the omniPAGE tank and lid allows the unit to be easily adapted for vertical PAGE or IEF with capillary tube gels, using the relevant optional insert.

  • Multi-transfer capability for 4 mini gels
  • Complete run-time flexibility from overnight to as short as a couple of hours
  • Cool pack for environmentally friendly low-cost cooling during high-intensity transfers
  • Open design, rigid cassettes maximise current transfer and eliminate shadow band formation
  • Cassettes colour coded prevent polarity reversal to ensure transfer in the correct orientation
Specifications
  • Description:
    Mini-wide blotting system
  • Dimensions:
    100×200 mm
  • Seq:
    02
  • Cat. No.:
    CLEVSB10W
  • Gel size W×L:
    160×85 mm
  • Buffer volume:
    2800 ml
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Fibre pads for 20×10 cm mini-wide blotter
    Fibre pads for 20×10 cm mini-wide blotter
    CLEVSB10WF
    Log in for availability
    1 * 6 items - £86.60

Product Information

  • Catalog #
    Availability
    Price
  • Compression cassette for 20×10 cm mini-wide blotter
    Compression cassette for 20×10 cm mini-wide blotter
    CLEVSB10WC
    Log in for availability
    1 * 1 items - £86.60

Product Information

  • Catalog #
    Availability
    Price
  • Mini-wide electroblotting module
    Mini-wide electroblotting module
    CLEVVS10WBI
    Log in for availability
    1 * 1 items - £754.00

Product Information

  • Catalog #
    Availability
    Price
  • Fibre pads for 20×20 cm maxi blotter
    Fibre pads for 20×20 cm maxi blotter
    CLEVSB20F
    Log in for availability
    1 * 4 items - £122.00

Product Information

  • Catalog #
    Availability
    Price
  • Maxi electroblotting module
    Maxi electroblotting module
    CLEVVS20BI
    Log in for availability
    1 * 1 items - £985.00

Product Information

  • Catalog #
    Availability
    Price
  • Fibre pads for 10×10 cm mini blotter
    Fibre pads for 10×10 cm mini blotter
    CLEVSB10F
    Log in for availability
    1 * 6 items - £72.70

Product Information

  • Catalog #
    Availability
    Price
  • Mini internal electroblotting module
    Mini internal electroblotting module
    CLEVVS10BI
    Log in for availability
    1 * 1 items - £661.00

Product Information

  • Catalog #
    Availability
    Price
  • Compression cassette for 20×20 cm maxi blotter
    Compression cassette for 20×20 cm maxi blotter
    CLEVSB20C
    Log in for availability
    1 * 1 items - £122.00

Product Information

  • Catalog #
    Availability
    Price
  • Compression cassette for 10×10 cm mini blotter
    Compression cassette for 10×10 cm mini blotter
    CLEVSB10C
    Log in for availability
    1 * 1 items - £72.70

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.