Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Chlorothioxanthone ≥99%

2-Chlorothioxanthone ≥99%

Glass bottle for solids, 5 g
Catalog # A18131.06
2-Chlorothioxanthone ≥99%
Catalog # A18131.06
CAS Number:  86-39-5
2-Chlorothioxanthone ≥99%
2-Chlorothioxanthone ≥99%

Glass bottle for solids, 5 g

CAS Number:  86-39-5
Price
£31.40
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 86-39-5 Molecular Formula: C13H7ClOS Formula Weight: 246.72 Flash Point: 91°(196°F) Melting Point: 151-154° MDL No.: MFCD00005067

Synonyms:  2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
  • CAS Number:86-39-5
  • Formula:C₁₃H₇ClOS
  • MDL Number:MFCD00005067
  • Molecular Weight:246.72 g/mol
  • Melting Pt:151…154 °C
  • Flash Pt:91 °C (196 °F)
  • Storage Temperature:Ambient
  • EINECS:201-667-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.