Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Brij® 35

Glass bottle for solids, 5.000 g
Catalog # A15809.0I
Irritant
Brij® 35
Catalog # A15809.0I
CAS Number:  9002-92-0
Brij® 35
Brij® 35

Glass bottle for solids, 5.000 g

CAS Number:  9002-92-0
Price
£212.00
Log in for availability
1 * 5.000 g
Product Details & Documents

CAS No.: 9002-92-0 Molecular Formula: C58H118O24 Formula Weight: 1199.57 Physical Form: Waxy solid Appearance: White Melting Point: ca 35-40° MDL No.: MFCD00080891

For use in Stein-Moore chromatography

Synonyms:  Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
  • CAS Number:9002-92-0
  • Formula:C₅₈H₁₁₈O₂₄
  • MDL Number:MFCD00080891
  • Molecular Weight:1199.56 g/mol
  • Melting Pt:∼ 35…40 °C
  • Storage Temperature:Ambient
  • EINECS:500-002-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications