Horizontal electrophoresis system, multiSUB mini
multiSUB mini 7, 70×70 mm, 2× 8-sample combs, loading guides and dams
Catalog # CLEVMSMINI7
Supplier: CLEAVER SCIENTIFIC
Jump to:
Product Details & Documents
The multiSUB mini is designed for low to medium numbers of samples. The small gel size maximises run economy but does not compromise versatility as two tray options are available – 70×70 mm and 70×100 mm – and combs ranging from preparative up to 16 samples. Simply by altering the gel tray or comb, this compact unit is capable of resolving up to 64 different samples, prepping 1 ml of sample or separating sample bands over a distance of 9 cm.
- Two tray options
- Run up to 64 samples
- Low buffer volumes
Specifications
- Description:multiSUB mini 7, 70×70 mm, 2× 8-sample combs, loading guides and dams
- Seq:01
- Cat. No.:CLEVMSMINI7
- Unit W×L×H:90×210×90 mm
- Gel size W×L:70×70 mm
- Sample capacity:32 samples
- Model:multiSUB mini 7
- Buffer volume:225 ml
Accessories
Product Information
- Catalog #AvailabilityPrice
- Negative electrode
Product Information
- Catalog #AvailabilityPrice
- Positive electrode
Product Information
- Catalog #AvailabilityPrice
- UV tray, 70×70 mm



