ASCARITE II CARBON DIOXIDE ABSORBENT, CA . 8-20 MESH 1 * 500 g
Catalog # SIAL223913-500G
Supplier: Merck
ASCARITE II CARBON DIOXIDE ABSORBENT, CA . 8-20 MESH 1 * 500 g
Catalog # SIAL223913-500G
Supplier: Merck
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® AVS TITRINORM, Silicon Standard Solution, 1000 mg/L Si in Water, Standard for AAS
Supplier: VWR Chemicals
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Diethylamine for synthesis
Supplier: Merck
Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
Supplier: Honeywell Chemicals