Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
Biopad biopsy pad, blue
Catalog # 720-2331
Biopsy specimen processing, CellSafe™
Catalog # 720-2331
Supplier:  
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
Biopad biopsy pad, blue
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Biopsy foam pad Q Path blue 1 * 500 items Catalog #720-2254
Product 720-2254 is similar to 720-2331 but may have a different manufacturer and/or packaging.
BIOPSY SPONGE 25.4 X 30.4 MM BLUE 1 * 1.000 items Catalog #720-0211
Product 720-0211 is similar to 720-2331 but may have a different manufacturer and/or packaging.
Product Details & Documents
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
  • No tissue damage
  • Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
Specifications
  • Description:
    Biopad biopsy pad, blue