Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Inoculating loops, platinum/iridium wire

Inoculating loops, platinum/iridium wire

Inoculating loop, platinum iridium wire, thickness 5 mm, 5 mm, 60 mm
Catalog # SCUT3686252
Supplier:   SCHUTT LABORTECHNIK
Inoculating loops, platinum/iridium wire
Catalog # SCUT3686252
Inoculating loops, platinum/iridium wire

Inoculating loop, platinum iridium wire, thickness 5 mm, 5 mm, 60 mm

Price
£52.10
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Ø:
    5 mm
  • Description:
    Inoculating loop, platinum iridium wire, thickness 5 mm
  • Length:
    60 mm
  • Seq:
    02