QuikChange Lightning Site-Directed 1 * 30 Tests
STRA210519
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific
Diethyl ether ≥99% stabilised for HPLC
: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
p-Toluidine, Supelco®
: Merck
Gentle Cell Dissociation Reagent
: STEMCELL Technologies