Density hydrometers, L50
L50, 0,800 - 0,850 g/cm³, 0,0005 g/cm³, 335 mm
AMARH806208/WC
: Amarell
Density hydrometers designed with or without verification certificate.
- Length:335 mm
- Seq:03
- Division:0,0005 g/cm³
- Model:L50
- Range:0,800 - 0,850 g/cm³



