Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Density hydrometers, L50

L50, 0,750 - 0,800 g/cm³, 0,0005 g/cm³, 335 mm
Catalog # AMARH806206/WC
Supplier:   Amarell
Density hydrometers, L50
Catalog # AMARH806206/WC
Supplier:  Amarell
Density hydrometers, L50
Density hydrometers, L50

L50, 0,750 - 0,800 g/cm³, 0,0005 g/cm³, 335 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Density hydrometers designed with or without verification certificate.

Specifications
  • Length:
    335 mm
  • Seq:
    02
  • Division:
    0,0005 g/cm³
  • Model:
    L50
  • Range:
    0,750 - 0,800 g/cm³
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.