PAHS IN TOLUENE (1.2ML) 1 * 5 items
PROONIST-2260A
Frequently Bought Together
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
5L Distilled Water 1 * 5 L
: REAGENT
Phenanthrene, Sigma-Aldrich®
: Merck
Clozapine N-oxide (CNO) (freebase), 50mg (>98%) 1 * 50 mg
: MARKET SOURCE PART PROCESS
2,2,4-Trimethylpentane 99.5% for analysis
: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Acetonylacetone, Sigma-Aldrich®
: Merck
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
: Honeywell Chemicals