2,5-Dichlorobenzaldehyde 98%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
2,5-Dichlorobenzaldehyde 98%
Catalog # ACRO359940010
Supplier: Thermo Fisher Scientific
CAS Number: 6361-23-5
Jump to:
Product Details & Documents
- CAS Number:6361-23-5
- Formula:C₇H₄Cl₂O
- MDL Number:MFCD00156140
- Molecular Weight:175.01 g/mol
- Melting Pt:54…58 °C
- EINECS:228-832-1
- UN:3261
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



