2,4-Dichlorophenylboronic acid 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
2,4-Dichlorophenylboronic acid 97%
Catalog # ACRO367670050
Supplier: Thermo Fisher Scientific
CAS Number: 68716-47-2
Jump to:
Product Details & Documents
Synonyms: Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid
- CAS Number:68716-47-2
- Formula:C₆H₅BCl₂O₂
- MDL Number:MFCD00013930
- Molecular Weight:190.82 g/mol
- Melting Pt:240…245 °C
Documents
ACRO36767
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



