ALKALINE-CYANIDE REAGENT 1 * 500 mL
Catalog # HACH2122349
Supplier: Hach
Frequently Bought Together
Chlorophyllin, coppered trisodium salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Methyl orange, Sigma-Aldrich®
Supplier: Merck
BIOTECTOR 1.8N SULFURIC ACID 80MG/L MN 1 * 20 L
Supplier: Hach
Plasma/Low Hb Control 5 g/l 1 * 1 items
Supplier: RADIOMETER CLINICAL PRODUCTS
2ML PL UREA 2 (50 TESTS) UREA PHOTOMETER 1 * 2 mL
Supplier: WATER ID PRIMELAB
QUANTI BLUE SOLUTION
Supplier: Invivogen
Silica monitor reagent (sodium molybdate) for seres FG 504B silicameter
Supplier: VWR Chemicals
1 component: Ethyl-tert-butylether (ETBE) [CAS:637-92-3] 2000mg/l in Met 1 * 5 Ampoul
Supplier: CUSTOM MADE CHEMICALS LAB
NEISSER´S SOLUTION III FOR MICROSCOPY 1 * 100 mL
Supplier: Roth Carl
Indigotrisulphonic acid potassium salt ≥60% (by HPLC) ozone-scavenging reagent, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY