MultiScreen® PCR 96 Filter Plates
Product Details & Documents
The polymerase chain reaction (PCR) is a highly effective fundamental technique in molecular biology. It offers a fast and efficient way to amplify specific DNA or RNA sequences from nucleic acids derived from different sources using enzymatic processes in a laboratory setting.
- Application: Sample preparation
- Binding type: Low binding surface
- Ambient
We have an extensive range of PCR consumables and equipment designed to facilitate every stage of your PCR workflow MultiScreen-PCR 96 Filter plates can be used to purify PCR products to be used in further downstream applications.
MultiScreen-PCR 96 filter plate is suitable for use:
-In receptor/ligand binding assays
-In sample preparation
-In enzyme assays with precipitation
-In DNA purification
-In polymerase chain reaction (PCR)
-In purifying polymerase chain reaction (PCR) products for mutation analysis
-In purifying PCR products for measuring CAG repeats
-In purifying PCR products for sequencing
Specifications
- Membrane:Proprietary
- No. of wells:96
- Recommended working volume:150 - 300 μl
- Sterile:-
- Description:MultiScreen PCR filter plate, with lid
- Material:Polystyrene
- Seq:01
- Well volume:300 μl



