Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

110 ml, 116 mm
Catalog # SMITC147/110
Supplier:   SMITH SCIENTIFIC
Büchner funnels
Catalog # SMITC147/110
Supplier:  SMITH SCIENTIFIC
Büchner funnels

110 ml, 116 mm

Price
£65.30
Log in for availability
1 * 1 items
Product Details & Documents

White glazed porcelain.

  • Very good chemical resistance
  • Very good chemical resistance
  • Easy to clean

Filtering funnels acc. to Büchner. For paper or membrane filtration discs.

Specifications
  • Ø:
    116 mm
  • Capacity:
    110 ml
  • Seq:
    03