Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

70 ml, 77 mm
Catalog # SMITC145/70
Supplier:   SMITH SCIENTIFIC
Büchner funnels
Catalog # SMITC145/70
Supplier:  SMITH SCIENTIFIC
Büchner funnels

70 ml, 77 mm

Price
£29.00
Log in for availability
1 * 1 items
Product Details & Documents

White glazed porcelain.

  • Very good chemical resistance
  • Very good chemical resistance
  • Easy to clean

Filtering funnels acc. to Büchner. For paper or membrane filtration discs.

Specifications
  • Ø:
    77 mm
  • Capacity:
    70 ml
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.