Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Soxhlet extractor
Soxhlet extractor
Soxhlet extractor body, 60 ml, borosilicate glass, 34/35 NS
Catalog # SMIT8SEC60/55
Supplier:   SMITH SCIENTIFIC
Soxhlet extractor
Catalog # SMIT8SEC60/55
Supplier:  SMITH SCIENTIFIC
Soxhlet extractor
Soxhlet extractor body, 60 ml, borosilicate glass, 34/35 NS
Price
£285.00
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass.
  • Excellent temperature resistance
  • Excellent chemical resistance
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Specifications
  • Socket size:
    34/35 NS
  • Description:
    Soxhlet extractor body, 60 ml, borosilicate glass
  • Seq:
    03