Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Filter discs
Filter discs
2, 30 mm
Catalog # SMIT11S2/30
Supplier:   SMITH SCIENTIFIC
Filter discs
Catalog # SMIT11S2/30
Supplier:  SMITH SCIENTIFIC
Filter discs
2, 30 mm
Price
£46.20
Log in for availability
1 * 1 items
Product Details & Documents
Sintered filter discs made of borosilicate glass.
Specifications
  • Ø:
    30 mm
  • Porosity:
    2
  • Seq:
    03