Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Test kits, chlorine, Spectroquant®, Supelco®
Chlorine reagent Cl₂-3*4, -, Prove, Nova 30/60, Move, Picco, Multy, 600 Tests
  1.00088.0001
 :   Merck
Test kits, chlorine, Spectroquant®, Supelco®
  1.00088.0001
 :  Merck
 :  
Test kits, chlorine, Spectroquant®, Supelco®
Test kits, chlorine, Spectroquant®, Supelco®
Chlorine reagent Cl₂-3*4, -, Prove, Nova 30/60, Move, Picco, Multy, 600 Tests
 :  
£64.60
1 * 600 Tests
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*4 Reagents for free chlorine testing are reagent Cl₂-1 1.00086.0001  and reagent Cl₂-2 1.00087.0001 together with empty cell and accessories  1.00089.0001
Reagents for total chlorine are reagent Cl₂-1 1.00086.0001, reagent Cl₂-2 1.00087.0001 and reagent Cl₂-3 1.00088.0001 together with empty cell and accessories 1.00089.0001
"*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and waste water
  • Pk:
    600 Tests
  • Description:
    Chlorine reagent Cl₂-3*<sup>4</sup>
  • For unit:
    Prove, Nova 30/60, Move, Picco, Multy
  • Range:
    -
  • Cat. No.:
    1.00088.0001
  • M-Clarity quality level:
    MQ100
Accessories
  •  
    Description
    For unit
    Pk
  • 1.00089.0001
    Chlorine, cells and accessories for photometric Cl₂ measurement*4
    Prove, Nova 60
    25
    1 * 25 items - £83.40