Cyclohexanone oxime ≥97%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Cyclohexanone oxime ≥97%
Catalog # A19820.36
Supplier: Thermo Fisher Scientific
CAS Number: 100-64-1
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 100-64-1 Molecular Formula: C6H11NO Formula Weight: 113.16 Flash Point: 103°(217°F) Melting Point: 87-91° Boiling Point: 206-210° Beilstein Reference: 1616769 MDL No.: MFCD00001660
- CAS Number:100-64-1
- Formula:C₆H₁₁NO
- MDL Number:MFCD00001660
- Molecular Weight:113.16 g/mol
- Boiling Pt:206…210 °C
- Melting Pt:87…91 °C
- Density:1.13 g/cm³ (20 °C)
- Flash Pt:103 °C (217 °F)
- Storage Temperature:Ambient
- EINECS:202-874-0
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Cat. No.:A19820.36
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



