Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cyclohexanone oxime ≥97%

Cyclohexanone oxime ≥97%

Glass bottle for solids, 100 g
Catalog # A19820.22
Toxic
Cyclohexanone oxime ≥97%
Catalog # A19820.22
CAS Number:  100-64-1
Cyclohexanone oxime ≥97%
Cyclohexanone oxime ≥97%

Glass bottle for solids, 100 g

CAS Number:  100-64-1
Price
£20.20
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 100-64-1 Molecular Formula: C6H11NO Formula Weight: 113.16 Flash Point: 103°(217°F) Melting Point: 87-91° Boiling Point: 206-210° Beilstein Reference: 1616769 MDL No.: MFCD00001660

  • CAS Number:100-64-1
  • Formula:C₆H₁₁NO
  • MDL Number:MFCD00001660
  • Molecular Weight:113.16 g/mol
  • Boiling Pt:206…210 °C
  • Melting Pt:87…91 °C
  • Density:1.13 g/cm³ (20 °C)
  • Flash Pt:103 °C (217 °F)
  • Storage Temperature:Ambient
  • EINECS:202-874-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications