Triethanolamine hydrochloride ≥99%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Triethanolamine hydrochloride ≥99%
Catalog # A15678.30
Supplier: Thermo Fisher Scientific
CAS Number: 637-39-8
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 637-39-8 Molecular Formula: C6H15NO3.HCl Formula Weight: 185.65 Melting Point: 177-179° Beilstein Reference: 3909940 Merck Reference: 14,9665 MDL No.: MFCD00012596
- CAS Number:637-39-8
- Formula:C₆H₁₆ClNO₃
- MDL Number:MFCD00012596
- Molecular Weight:185.65 g/mol
- Merck Index:14,9665
- Melting Pt:177…179 °C
- Storage Temperature:Ambient
- EINECS:211-284-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



