Phthalide ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 87-41-2 Molecular Formula: C8H6O2 Formula Weight: 134.13 Flash Point: 152°(305°F) Melting Point: 72-75° Boiling Point: 289-291° Beilstein Reference: 114632 MDL No.: MFCD00005906
Synonyms: Isobenzofuran-1(3H)-one
- CAS Number:87-41-2
- Formula:C₈H₆O₂
- MDL Number:MFCD00005906
- Molecular Weight:134.13 g/mol
- Boiling Pt:289…291 °C
- Melting Pt:72…75 °C
- Density:1.1
- Flash Pt:152 °C (305 °F)
- Storage Temperature:Ambient
- EINECS:201-744-0
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



