Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

3-Pentanone ≥99%

Glass bottle, 500 mL
Catalog # A15297.AP
Flammable
Irritant
3-Pentanone ≥99%
Catalog # A15297.AP
CAS Number:  96-22-0
3-Pentanone ≥99%
3-Pentanone ≥99%

Glass bottle, 500 mL

CAS Number:  96-22-0
Price
£29.60
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 96-22-0 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(45°F) Refractive Index: 1.3920 Odour: Acetone-like Melting Point: -39° Boiling Point: 102-103° Beilstein Reference: 635749 Merck Reference: 14,3121 UN No.: 1156 MDL No.: MFCD00009320

Synonyms:  Diethyl ketone
  • CAS Number:96-22-0
  • Formula:C₅H₁₀O
  • MDL Number:MFCD00009320
  • Molecular Weight:86.13 g/mol
  • Merck Index:14,3121
  • Boiling Pt:102…103 °C
  • Melting Pt:–39 °C
  • Density:0.812
  • Flash Pt:7 °C (45 °F)
  • Storage Temperature:Ambient
  • EINECS:202-490-3
  • UN:1156
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications