Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
3-Pentanone ≥99%

3-Pentanone ≥99%

Glass bottle, 100 mL
Catalog # A15297.AE
Flammable
Irritant
3-Pentanone ≥99%
Catalog # A15297.AE
CAS Number:  96-22-0
3-Pentanone ≥99%
3-Pentanone ≥99%

Glass bottle, 100 mL

CAS Number:  96-22-0
Price
£24.80
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99% CAS No.: 96-22-0 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(45°F) Refractive Index: 1.3920 Odour: Acetone-like Melting Point: -39° Boiling Point: 102-103° Beilstein Reference: 635749 Merck Reference: 14,3121 UN No.: 1156 MDL No.: MFCD00009320

Synonyms:  Diethyl ketone
  • CAS Number:96-22-0
  • Formula:C₅H₁₀O
  • MDL Number:MFCD00009320
  • Molecular Weight:86.13 g/mol
  • Merck Index:14,3121
  • Boiling Pt:102…103 °C
  • Melting Pt:–39 °C
  • Density:0.812
  • Flash Pt:7 °C (45 °F)
  • Storage Temperature:Ambient
  • EINECS:202-490-3
  • UN:1156
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.