Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

4,4'-Dihydroxybenzophenone ≥98%

Glass bottle for solids, 100 g
Catalog # A15097.22
Irritant
4,4'-Dihydroxybenzophenone ≥98%
Catalog # A15097.22
CAS Number:  611-99-4
4,4'-Dihydroxybenzophenone ≥98%
4,4'-Dihydroxybenzophenone ≥98%

Glass bottle for solids, 100 g

CAS Number:  611-99-4
Price
£268.00
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 611-99-4 Molecular Formula: C13H10O3 Formula Weight: 214.22 Melting Point: 221-225° Beilstein Reference: 1874572

Synonyms:  Bis(4-hydroxyphenyl) ketone
  • CAS Number:611-99-4
  • Formula:C₁₃H₁₀O₃
  • MDL Number:MFCD00002358
  • Molecular Weight:214.22 g/mol
  • Boiling Pt:375 °C (1013 hPa)
  • Melting Pt:221…225 °C
  • Density:1.336 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:210-288-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications