2-Fluorophenol ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 367-12-4 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 46°(114°F) Refractive Index: 1.5140 Odour: Phenol-like Melting Point: 14-16° Boiling Point: 150-152° Beilstein Reference: 1905112 UN No.: 2924 MDL No.: MFCD00002155
Synonyms: o-Fluorophenol
- CAS Number:367-12-4
- Formula:C₆H₅FO
- MDL Number:MFCD00002155
- Molecular Weight:112.1 g/mol
- Boiling Pt:150…152 °C
- Melting Pt:14…16 °C
- Density:1.215
- Flash Pt:46 °C (114 °F)
- Storage Temperature:Ambient
- EINECS:206-681-2
- UN:2924
- ADR:3,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Cat. No.:A14979.22
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



