Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4-Chlorophenyl isocyanate ≥98%

4-Chlorophenyl isocyanate ≥98%

Glass bottle for solids, 5 g
Catalog # A14966.06
Toxic
Health
Corrosive
Irritant
4-Chlorophenyl isocyanate ≥98%
Catalog # A14966.06
CAS Number:  104-12-1
4-Chlorophenyl isocyanate ≥98%
4-Chlorophenyl isocyanate ≥98%

Glass bottle for solids, 5 g

CAS Number:  104-12-1
Price
£21.50
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 104-12-1 Molecular Formula: C7H4ClNO Formula Weight: 153.57 Storage Temperature: 2°C to 8°C Flash Point: 90°(194°F) Refractive Index: 1.5618 Odour: Pungent Melting Point: 29-31° Boiling Point: 203-204° Beilstein Reference: 386235 UN No.: 2811 MDL No.: MFCD00002024

  • CAS Number:104-12-1
  • Formula:C₇H₄ClNO
  • MDL Number:MFCD00002024
  • Molecular Weight:153.57 g/mol
  • Boiling Pt:203…204 °C
  • Melting Pt:29…31 °C
  • Density:1.200
  • Flash Pt:90 °C (194 °F)
  • Storage Temperature:Refrigerator
  • EINECS:203-176-9
  • ADR:6.1,II
  •  :Density: 1.2 g/cm³ (20 °C)
  • UN:2811
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications