Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cyclohexanecarboxylic acid ≥98%

Cyclohexanecarboxylic acid ≥98%

Glass bottle for solids, 500 g
Catalog # A14693.36
Irritant
Cyclohexanecarboxylic acid ≥98%
Catalog # A14693.36
CAS Number:  98-89-5
Cyclohexanecarboxylic acid ≥98%
Cyclohexanecarboxylic acid ≥98%

Glass bottle for solids, 500 g

CAS Number:  98-89-5
Price
£51.50
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 98-89-5 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 118°(244°F) Refractive Index: 1.4610 Melting Point: 29-32° Boiling Point: 231-233° Beilstein Reference: 970529 Merck Reference: 14,2724 MDL No.: MFCD00001461

Synonyms:  Hexahydrobenzoic acid
  • CAS Number:98-89-5
  • Formula:C₇H₁₂O₂
  • MDL Number:MFCD00001461
  • Molecular Weight:128.17 g/mol
  • Merck Index:14,2724
  • Boiling Pt:231…233 °C
  • Melting Pt:29…32 °C
  • Density:1.048
  • Flash Pt:118 °C (244 °F)
  • Storage Temperature:Ambient
  • EINECS:202-711-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications