Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Iodophenol ≥98%
Glass bottle for solids, 100 g
  A13599.22
2-Iodophenol ≥98%
  A13599.22
 :  533-58-4
2-Iodophenol ≥98%
2-Iodophenol ≥98%
Glass bottle for solids, 100 g
 :  533-58-4
£166.00
1 * 100 g
Purity: 98% CAS No.: 533-58-4 Molecular Formula: C6H5IO Formula Weight: 220.01 Flash Point: 110°(230°F) Melting Point: 39-41° Boiling Point: 186-187°/160mm Beilstein Reference: 1855300 Merck Reference: 14,5035 MDL No.: MFCD00013963
  •  :533-58-4
  •  :C₆H₅IO
  •  :MFCD00013963
  •  :220.01 g/mol
  •  :14,5035
  •  :186…187 °C (160 mmHg)
  •  :39…41 °C
  •  :1.947
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :208-569-9
  •  :2811
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g