Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sodium-2-ethylhexanoate ≥97%
Glass bottle for solids, 250 g
  B24960.30
Sodium-2-ethylhexanoate ≥97%
  B24960.30
 :  19766-89-3
Sodium-2-ethylhexanoate ≥97%
Sodium-2-ethylhexanoate ≥97%
Glass bottle for solids, 250 g
 :  19766-89-3
£26.40
1 * 250 g
Purity: 97% CAS No.: 19766-89-3 Molecular Formula: C8H15NaO2 Formula Weight: 166.20 Odour: Odourless Melting Point: 300° MDL No.: MFCD00014007
 :  2-Ethylcaproic acid sodium salt, Sodium 2-ethylhexanoate, 2-Ethylhexanoic acid sodium salt
  •  :19766-89-3
  •  :C₈H₁₅NaO₂
  •  :MFCD00014007
  •  :166.2 g/mol
  •  :>300 °C
  •  :Ambient
  •  :243-283-8
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g