Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Sodium-2-ethylhexanoate ≥97%

Glass bottle for solids, 5.000 g
Catalog # B24960.0I
Health
Sodium-2-ethylhexanoate ≥97%
Catalog # B24960.0I
CAS Number:  19766-89-3
Sodium-2-ethylhexanoate ≥97%
Sodium-2-ethylhexanoate ≥97%

Glass bottle for solids, 5.000 g

CAS Number:  19766-89-3
Price
£268.00
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 97% CAS No.: 19766-89-3 Molecular Formula: C8H15NaO2 Formula Weight: 166.20 Odour: Odourless Melting Point: 300° MDL No.: MFCD00014007

Synonyms:  2-Ethylcaproic acid sodium salt, Sodium 2-ethylhexanoate, 2-Ethylhexanoic acid sodium salt
  • CAS Number:19766-89-3
  • Formula:C₈H₁₅NaO₂
  • MDL Number:MFCD00014007
  • Molecular Weight:166.2 g/mol
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:243-283-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications