Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4-Bromo-2-chlorophenol ≥98%

4-Bromo-2-chlorophenol ≥98%

Glass bottle for solids, 50 g
Catalog # B24803.18
Irritant
4-Bromo-2-chlorophenol ≥98%
Catalog # B24803.18
CAS Number:  3964-56-5
4-Bromo-2-chlorophenol ≥98%
4-Bromo-2-chlorophenol ≥98%

Glass bottle for solids, 50 g

CAS Number:  3964-56-5
Price
£60.40
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 98% CAS No.: 3964-56-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Flash Point: 110°(230°F) Odour: Phenol-like Boiling Point: 232-235° Beilstein Reference: 2042867 MDL No.: MFCD00002166

  • CAS Number:3964-56-5
  • Formula:C₆H₄BrClO
  • MDL Number:MFCD00002166
  • Molecular Weight:207.45 g/mol
  • Boiling Pt:232…235 °C
  • Melting Pt:47…50 °C
  • Density:1.617 g/cm³ (20 °C)
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:223-572-5
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.