Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Congo red indicator

Congo red indicator

100 g
Catalog # B24310.22
Health
Congo red indicator
Catalog # B24310.22
CAS Number:  573-58-0
Congo red indicator
Congo red indicator

100 g

CAS Number:  573-58-0
Price
£96.50
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Formula Weight: 696.67 Odour: Odourless Melting Point: 360° Beilstein Reference: 3894858 Merck Reference: 14,2498 MDL No.: MFCD00004028

Synonyms:  Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), CI 22120, Direct Red 28
  • CAS Number:573-58-0
  • Formula:C₃₂H₂₂N₆Na₂O₆S₂
  • MDL Number:MFCD00004028
  • Molecular Weight:696.67 g/mol
  • Merck Index:14,2498
  • Melting Pt:>360 °C
  • Storage Temperature:Ambient
  • EINECS:209-358-4
Specifications

Specifications

  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.