Congo red indicator
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Formula Weight: 696.67 Odour: Odourless Melting Point: 360° Beilstein Reference: 3894858 Merck Reference: 14,2498 MDL No.: MFCD00004028
Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), CI 22120, Direct Red 28
- CAS Number:573-58-0
- Formula:C₃₂H₂₂N₆Na₂O₆S₂
- MDL Number:MFCD00004028
- Molecular Weight:696.67 g/mol
- Merck Index:14,2498
- Melting Pt:>360 °C
- Storage Temperature:Ambient
- EINECS:209-358-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



