Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Hexyne ≥98%

2-Hexyne ≥98%

Glass bottle for solids, 10 g
Catalog # B22405.09
Flammable
Health
Irritant
2-Hexyne ≥98%
Catalog # B22405.09
CAS Number:  764-35-2
2-Hexyne ≥98%
2-Hexyne ≥98%

Glass bottle for solids, 10 g

CAS Number:  764-35-2
Price
£41.30
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 98+% CAS No.: 764-35-2 Molecular Formula: CH3(CH2)2C≡CCH3 Formula Weight: 82.15 Flash Point: -10°(14°F) Refractive Index: 1.4140 Odour: Characteristic Melting Point: -90° Boiling Point: 84-86° Beilstein Reference: 1731542 UN No.: 3295 MDL No.: MFCD00009470

  • CAS Number:764-35-2
  • Formula:C₆H₁₀
  • MDL Number:MFCD00009470
  • Molecular Weight:82.15 g/mol
  • Boiling Pt:84…86 °C
  • Melting Pt:–90 °C
  • Density:0.736
  • Flash Pt:–10 °C (14 °F)
  • Storage Temperature:Ambient
  • EINECS:212-117-6
  • UN:3295
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.