9-Fluorenone ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 486-25-9 Molecular Formula: C13H8O Formula Weight: 180.21 Flash Point: 163°(325°F) Physical Form: Cryst powder or flakes Density: 0.9 Appearance: Yellow Melting Point: 82-85° Boiling Point: 342° Beilstein Reference: 1636531 MDL No.: MFCD00001141
Synonyms: 9H-Fluoren-9-one
- CAS Number:486-25-9
- Formula:C₁₃H₈O
- MDL Number:MFCD00001141
- Molecular Weight:180.21 g/mol
- Boiling Pt:342 °C
- Melting Pt:82…85 °C
- Density:0.9
- Flash Pt:163 °C (325 °F)
- Storage Temperature:Ambient
- EINECS:207-630-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



