9-Fluorenone ≥98%
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 486-25-9 Molecular Formula: C13H8O Formula Weight: 180.21 Flash Point: 163°(325°F) Physical Form: Cryst powder or flakes Density: 0.9 Appearance: Yellow Melting Point: 82-85° Boiling Point: 342° Beilstein Reference: 1636531 MDL No.: MFCD00001141
Synonyms: 9H-Fluoren-9-one
- CAS Number:486-25-9
- Formula:C₁₃H₈O
- MDL Number:MFCD00001141
- Molecular Weight:180.21 g/mol
- Boiling Pt:342 °C
- Melting Pt:82…85 °C
- Density:0.9
- Flash Pt:163 °C (325 °F)
- Storage Temperature:Ambient
- EINECS:207-630-7
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



