Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4'-Hydroxyacetophenone ≥99%

4'-Hydroxyacetophenone ≥99%

Glass bottle for solids, 500 g
Catalog # A11974.36
Toxic
Irritant
4'-Hydroxyacetophenone ≥99%
Catalog # A11974.36
CAS Number:  99-93-4
4'-Hydroxyacetophenone ≥99%
4'-Hydroxyacetophenone ≥99%

Glass bottle for solids, 500 g

CAS Number:  99-93-4
Price
£133.00
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 99-93-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 166°(330°F) Melting Point: 106-108° Boiling Point: 286° Beilstein Reference: 774355 MDL No.: MFCD00002359

  • CAS Number:99-93-4
  • Formula:C₈H₈O₂
  • MDL Number:MFCD00002359
  • Molecular Weight:136.15 g/mol
  • Boiling Pt:286 °C
  • Melting Pt:106…108 °C
  • Density:1.139 g/cm³ (25 °C)
  • Flash Pt:166 °C (330 °F)
  • Storage Temperature:Ambient
  • EINECS:202-802-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.