Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4'-Hydroxyacetophenone ≥99%

4'-Hydroxyacetophenone ≥99%

Glass bottle for solids, 100 g
Catalog # A11974.22
Toxic
Irritant
4'-Hydroxyacetophenone ≥99%
Catalog # A11974.22
CAS Number:  99-93-4
4'-Hydroxyacetophenone ≥99%
4'-Hydroxyacetophenone ≥99%

Glass bottle for solids, 100 g

CAS Number:  99-93-4
Price
£46.50
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 99-93-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 166°(330°F) Melting Point: 106-108° Boiling Point: 286° Beilstein Reference: 774355 MDL No.: MFCD00002359

  • CAS Number:99-93-4
  • Formula:C₈H₈O₂
  • MDL Number:MFCD00002359
  • Molecular Weight:136.15 g/mol
  • Boiling Pt:286 °C
  • Melting Pt:106…108 °C
  • Density:1.139 g/cm³ (25 °C)
  • Flash Pt:166 °C (330 °F)
  • Storage Temperature:Ambient
  • EINECS:202-802-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications