Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4'-Hydroxyacetophenone ≥99%
Glass bottle for solids, 1.000 g
  A11974.0B
4'-Hydroxyacetophenone ≥99%
  A11974.0B
 :  99-93-4
4'-Hydroxyacetophenone ≥99%
4'-Hydroxyacetophenone ≥99%
Glass bottle for solids, 1.000 g
 :  99-93-4
£197.00
1 * 1.000 g
Purity: 99% CAS No.: 99-93-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 166°(330°F) Melting Point: 106-108° Boiling Point: 286° Beilstein Reference: 774355 MDL No.: MFCD00002359
  •  :99-93-4
  •  :C₈H₈O₂
  •  :MFCD00002359
  •  :136.15 g/mol
  •  :286 °C
  •  :106…108 °C
  •  :1.139 g/cm³ (25 °C)
  •  :166 °C (330 °F)
  •  :Ambient
  •  :202-802-8
  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g