Cation exchange chromatography columns, TSK-GEL® SP-5PW
Preparative guardgel kit, Guard cartridge, SP-5PW, -, 20 µm
Catalog # TOYO0016093
Supplier: TOSOH Bioscience
Cation exchange chromatography columns, TSK-GEL® SP-5PW
Catalog # TOYO0016093
Supplier: TOSOH Bioscience
Jump to:
Product Details & Documents
TSK-GEL® SP-5PW are polymer based column packed with porous hydrophilic polymer beads which are surface modified with a strong cation exchanger. Used for the separation and analysis of proteins, peptides, and other biologically active molecules.
Specifications
- Column Format:Guard cartridge
- Particle Size:20 µm
- Pore size:-
- Description:Preparative guardgel kit
- Seq:09
- Phase:SP-5PW