Order Entry
United Kingdom
ContactUsLinkComponent
Tst Control Strips Pk100 121Oc - 15 Minutes 1 * 100 items
Catalog # STER2342AB
Supplier:  STERIS
Tst Control Strips Pk100 121Oc - 15 Minutes 1 * 100 items
Catalog # STER2342AB
Supplier:  STERIS
Frequently Bought Together